You Searched For: Aquarium Tanks
Clear soda glass aquarium tanks are strong and thick-walled, suitable for a large range of applications. Their ground rims are smooth to the touch. These glass enclosures are ideal for applications without large demands on temperature stability, and sturdy walls will withstand regular use for many years. Clear sides allow for unobstructed visual inspection of submerged samples such as chromatography plates. Bottoms are ground to provide a flat, level surface.
Clear soda glass aquarium tanks are strong and thick-walled, suitable for a large range of applications. Their ground rims are smooth to the touch. These glass enclosures are ideal for applications without large demands on temperature stability, and sturdy walls will withstand regular use for many years. Clear sides allow for unobstructed visual inspection of submerged samples such as chromatography plates. Bottoms are ground to provide a flat, level surface.
4  results were found
SearchResultCount:"4"
Sort Results
List View Easy View
Rate These Search Results
Supplier: GLASWARENFABRIK KARL HECHT
Description: Tanks made from soda-lime glass for multipurpose use.
Supplier: MARIENFELD
Description: Aquarium tanks for a large range of applications. Made from clear soda glass.
Supplier: NEUBERT VOLUM. GLASSWAERE
Description: Soda-lime glass.
Supplier: NEUBERT VOLUM. GLASSWAERE
Description: Soda-lime glass
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
Product(s) marked with this symbol are discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.
1 - 4 of 4
no targeter for Bottom