Cryotubes Stor-It™, Nunc™
Supplier: Thermo Fisher Scientific
Nunc™ Stor-It™
391-8877EA 0 EUR
391-8877
Cryotubes Stor-It™, Nunc™
Vials
PP cryotubes with PE screw cap.
  • Free standing tubes with starfoot
  • External thread
  • With writing area
Packaging: Caps and vials packed separately.
Caution: Not recommended for use in the liquid phase of nitrogen.