You Searched For: Sample Concentrators
Sample concentrators for use with block heaters quickly evaporate liquid components for easy sample concentration when utilized with block heaters. Stainless steel needles deliver gas over the sample surface, producing quicker results than heating alone. The needles are available in various sizes to accommodate different tube lengths. Sample concentrators feature an adjustable stand, offering simple height control and flexibility to work with any content required.
Sample concentrators for use with block heaters quickly evaporate liquid components for easy sample concentration when utilized with block heaters. Stainless steel needles deliver gas over the sample surface, producing quicker results than heating alone. The needles are available in various sizes to accommodate different tube lengths. Sample concentrators feature an adjustable stand, offering simple height control and flexibility to work with any content required.
2  results were found
SearchResultCount:"2"
Sort Results
List View Easy View
Rate These Search Results
Supplier: Cole-Parmer
Description: Quickly concentrate multiple samples in a block at once.
Catalog Number: (HYPETS-18826)
Supplier: Thermo Fisher Scientific
Description: Reacti-Vap Evaporators (9 port)
These evaporators are precision machined gassing manifolds. They provide simple, efficient evaporation by ...
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
Product(s) marked with this symbol are discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.
1 - 2 of 2
no targeter for Bottom