You Searched For: Disinfection Systems
Disinfection systems come in a wide variety of shapes and processes, and the benefits of each are thorough and safe microbiological protection. These options include chlorine dioxide systems, electrolysis systems, ozone systems, and UV systems. UV systems work entirely without chemicals, as UV radiation alone produces microbiologically safe water with no harmful disinfection by-products. Ozone is the strongest oxidant and disinfectant currently used in water treatment, and it is extremely effective against bacteria and viruses.
Disinfection systems come in a wide variety of shapes and processes, and the benefits of each are thorough and safe microbiological protection. These options include chlorine dioxide systems, electrolysis systems, ozone systems, and UV systems. UV systems work entirely without chemicals, as UV radiation alone produces microbiologically safe water with no harmful disinfection by-products. Ozone is the strongest oxidant and disinfectant currently used in water treatment, and it is extremely effective against bacteria and viruses.
2  results were found
SearchResultCount:"2"
Sort Results
List View Easy View
Rate These Search Results
Catalog Number: (141-1546)
Supplier: Diversey
Description: MoonBeamâ„¢3 Disinfection Technology is a UV-C device that provides an additional reduction of contamination on pre-cleaned and disinfected medical devi...
UOM: 1 * 1 items
Supplier: BASAN
Description: This airborne disinfection system is based on a patented technology using a rotating disk to deliver an active agent in the form of microscopic drople...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
Product(s) marked with this symbol are discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.
1 - 2 of 2
no targeter for Bottom