VWR® kuroGEL, Electroblotting Modules For Verti Dual Gel Units
Supplier: VWR
VWR®
700-7283EA 754 EUR
700-7283
VWR® kuroGEL, Electroblotting Modules For Verti Dual Gel Units
Blotting Apparatus
These electroblotters are designed to provide a safe and easy to operate system for the transfer of nucleic acid or proteins from a gel matrix onto a suitable transfer membrane. Two sizes of unit can be used to blot up to two gels of either 10×10 cm or 20×20 cm.
  • Each asymmetric, colour coded compression cassette has a locking mechanism to minimise gel movement during insertion into the tank
  • The asymmetry of the cassettes prevents them being inserted into the tank in the wrong orientation, against the direction of transfer
Delivery information: Each 10×10 cm module is supplied with two 10 cm compression cassettes and two small fibre pads. Each 20×20 cm module is supplied with two 20 cm compression cassettes and two large fibre pads.