You Searched For: Vertical Gel Electrophoresis Systems

Vertical gel electrophoresis systems are primarily used for nucleic acid or protein sequencing. Achieve precise voltage control using this format that forces charged molecules to travel through the casted gel since it is the only buffer chamber connection. The low current used with the vertical gel systems does not require buffer to be recirculated. With strong clamps to guarantee the assembly stays in an upright position, the equipment facilitates fast gel runs with even cooling resulting in distinct bands.

Vertical gel electrophoresis systems are primarily used for nucleic acid or protein sequencing. Achieve precise voltage control using this format that forces charged molecules to travel through the casted gel since it is the only buffer chamber connection. The low current used with the vertical gel systems does not require buffer to be recirculated. With strong clamps to guarantee the assembly stays in an upright position, the equipment facilitates fast gel runs with even cooling resulting in distinct bands.


20  results were found

SearchResultCount:"20"

Sort Results

List View Easy View

Rate These Search Results

Supplier: VWR Collection
Description: The PerfectBlue Twin S system is designed for rapid separation of typical 10×10 cm mini gels. Due to its small size and a separation capacity of 6 to ...

Supplier: VWR Collection
Description: The shiroGEL vertical 20 offers application flexibility and is distinguished by its ease of gel casting and set-up, requiring only four screws to secu...

Supplier: VWR Collection
Description: Extra wide, for gels 20×10 cm, Twin ExW S is ideally suited to rapid analytical separations or large sample separation. The combination of the triple-...

Supplier: VWR Collection
Description: The largest of the series, the Twin L can accommodate 20×20 cm gels for the simultaneous separation of up to 50 samples making this system ideally sui...

Supplier: VWR Collection
Description: For 16×14 cm gels, the Twin M is a versatile system for SDS PAGE applications, in addition to non denaturing electrophoresis of proteins, SSCPs and di...

Catalog Number: (700-0292)
Supplier: VWR Collection
Description: The shiroGEL mini vertical PAGE system is modular, allowing PAGE and electroblotting to be carried out in the same tank, simply by changing the insert...
UOM: 1 * 1 items


Catalog Number: (730-0311)
Supplier: OWL SCIENTIFIC
Description: The Owlâ„¢ P81 is a compact, single sided vertical electrophoresis system designed for economical, quick protein runs.
UOM: 1 * 1 items


Supplier: OWL SCIENTIFIC
Description: The Owlâ„¢ P9DS dual gel system is designed for quick and easy casting and running of one or two gels on the same device. When used with the available c...

Catalog Number: (732-1923)
Supplier: HOEFER ELECTROPHORESIS
Description: Vertical gel electrophoresis system
UOM: 1 * 1 items


Catalog Number: (720-2856)
Supplier: OWL SCIENTIFIC
Description: Vertical gel electrophoresis system, Dual gel vertical gel electrophoresis, Dual gel, uncooled, Owl P82, 100×80 - 100 mm
UOM: 1 * 1 items


Catalog Number: (80-6418-77)
Supplier: Cytiva
Description: The simple, modular design of miniVE vertical electrophoresis system consolidates all the small parts for electrophoresis and blotting into one conven...
UOM: 1 * 1 items


Catalog Number: (732-1427)
Supplier: EDVOTEK EUROPE LTD
Description: Designed for separation of proteins on polyacrylamide gels, the MV10 unit holds one 9×19 cm gel cassette and can accommodate most pre-cast or self-mad...
UOM: 1 * 1 items


Catalog Number: (80-6479-57)
Supplier: Cytiva
Description: The key to outstanding, reproducible electrophoresis results is temperature control. Gels unequally cooled on their two faces produce slanted bands ap...
UOM: 1 * 1 items


Supplier: CLEAVER SCIENTIFIC
Description: Gradient mixer is ideal for caesium, sucrose and gel gradients.

Supplier: CLEAVER SCIENTIFIC
Description: By combining both functionality and ease of use, the OmniPAGE CVS10 systems provide complete flexibility for individual research needs. They can accom...

Supplier: CLEAVER SCIENTIFIC
Description: omniPAGE mini wide vertical gel unit with an active gel width of 18 cm effectively allows double the number of samples to be resolved as the omniPAGE ...

Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
Product(s) marked with this symbol are discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.
1 - 16 of 20
no targeter for Bottom