You Searched For: Rotary Evaporators
Rotary evaporators remove volatile solvents from non-volatile reaction samples through evaporation. The overall solvent removal rate is increased by reducing the pressure and rotating for even heat application to surface area. This is not only the quickest method, but it eliminates the chance of overheating as the solvent is collected into the flask. The rotary evaporators feature adjustable knobs for precise apparatus positioning, rotation speed, and temperature of water bath.
Rotary evaporators remove volatile solvents from non-volatile reaction samples through evaporation. The overall solvent removal rate is increased by reducing the pressure and rotating for even heat application to surface area. This is not only the quickest method, but it eliminates the chance of overheating as the solvent is collected into the flask. The rotary evaporators feature adjustable knobs for precise apparatus positioning, rotation speed, and temperature of water bath.
12  results were found
SearchResultCount:"12"
Sort Results
List View Easy View
Rate These Search Results
Supplier: VWR Collection
Description: This is a basic chemical laboratory instrument suitable for evaporation, distillation and separation of chemicals. It can be used as a multifunctional...
Sale
Supplier: VWR Collection
Description: This is an essential instrument for use in chemical laboratories for efficient and gentle removal of solvents from samples by evaporation. It offers a...
Supplier: Heidolph Instruments GmbH & Co.KG
Description: These evaporators are equipped with proven smart details such as Easy-Clip, patented clamping sleeve, lift stop to limit immersion depth and universal...
Supplier: Heidolph Instruments GmbH & Co.KG
Description: Hei-VAP Expert Control rotary evaporators allow central control of all process parameters: Vacuum, cooling temperature, rotation and heating bath temp...
Supplier: IKA
Description: Rotary evaporators with integrated heating bath, features include an innovative control concept with a bright, high-contrast, TFT, colour-graphic cont...
Supplier: IKA
Description: These entry-level evaporators are ideal for all standard evaporation applications in universities and research laboratories. The functions of the RV10...
Supplier: IKA
Description: Rotary evaporators with integrated heating bath, safe and simple operation via ergonomic controls situated at the front.
Supplier: IKA
Description: Rotary evaporators with integrated heating bath and vacuum controller; units feature USB interface, central display for automatic distillation and ram...
Supplier: IKA
Description: The RV 3 rotary evaporator is the ideal entry-level model from the IKA rotary evaporator portfolio. It is suitable for a multitude of uses in the chem...
Catalog Number: (531-1454)
Supplier: IKA
Description: The RV 10 auto pro V-C Complete package is the complete solution for your rotary evaporator set-up. In addition to the rotary evaporator RV 10 auto, t...
UOM: 1 * 1 items
Supplier: Cole Parmer
Description: Quality, efficient evaporators at an affordable price.
Catalog Number: (BUCH11I300S40)
Supplier: BUCHI
Description: Interface I-300 for use with BUCHI Rotavapour® R-21x/V-7x0 models.
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
Product(s) marked with this symbol are discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.
1 - 12 of 12
no targeter for Bottom