You Searched For: HPLC/GPC Pumps
The purpose of HPLC pumps is to maintain a constant flow of mobile phase through the HPLC regardless of the pressure caused by the flow resistance of the packed column. Most commercial HPLC pumps are based on a reciprocating piston design. HPLC pumps can be used to mix or blend the mobile phase as well as to deliver it to the system. Their function is to reduce separation time of large column packings and allow the use of smaller particle size packings.
The purpose of HPLC pumps is to maintain a constant flow of mobile phase through the HPLC regardless of the pressure caused by the flow resistance of the packed column. Most commercial HPLC pumps are based on a reciprocating piston design. HPLC pumps can be used to mix or blend the mobile phase as well as to deliver it to the system. Their function is to reduce separation time of large column packings and allow the use of smaller particle size packings.
3  results were found
SearchResultCount:"3"
Sort Results
List View Easy View
Rate These Search Results
Catalog Number: (GILI38114001)
Supplier: GILSON LCMS
Description: Ideal for enabling chemical reaction monitoring, and supporting systems for environmental, food and beverage, and petroleum applications and for gel p...
UOM: 1 * 1 items
Supplier: Avantor Fluid Handling
Description: Designed for optimal operation in your Ismatec pumps.
Supplier: Avantor Fluid Handling
Description: Remote flow rate control via analog signal or RS232 interface.
CE Compliant
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
Product(s) marked with this symbol are discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.
1 - 3 of 3
no targeter for Bottom