VWR® VDI 12, Homogeniser
Supplier: VWR
VWR®
431-0125EA 1130 EUR
431-0125 431-0124 431-0109
VWR® VDI 12, Homogeniser
Homogenisers Homogenisers Handheld
Handheld or stand model, volume range 0,1 to 100 ml. An ergonomic, lightweight, handheld homogeniser which is easy to handle. Ideal for RNA protein assays, DNA and RNA blot analysis, PCR work, wastewater extractions, tissue decomposition and suspensions. High shear homogenisation reduces sample process times 10-fold compared to conventional mixers.
  • Motor accepts VWR® dispersing elements as well as those from selected other manufacturers*
  • Overload protection means motor shuts down if overload conditions arise, ensuring the motor is protected
  • Quick release coupling enables dispersing elements to be changed quickly and easily
Certifications: IP protection class according to DIN EN 60529: IP 30
Ordering information: Supplied without dispersing elements, please order required accessories separately.
Caution: * For details of dispersing elements please contact Avantor.