You Searched For: UV Viewing Cabinets

The ultraviolet viewing cabinet have specialized viewports for contrast observation that even accommodate users that wear prescription glasses. Filters to protect the eyes from harmful shortwave radiation and block the blue haze associated with longwave UV radiation. All UV viewing cabinet models feature removable bottom floor panels for placement over large samples or transilluminators. The lightweight access curtains allow for easy entry and prohibit any external light.

The ultraviolet viewing cabinet have specialized viewports for contrast observation that even accommodate users that wear prescription glasses. Filters to protect the eyes from harmful shortwave radiation and block the blue haze associated with longwave UV radiation. All UV viewing cabinet models feature removable bottom floor panels for placement over large samples or transilluminators. The lightweight access curtains allow for easy entry and prohibit any external light.


5  results were found

SearchResultCount:"5"

Sort Results

List View Easy View

Rate These Search Results

Supplier: UVP ULTRA VIOLET PRODUCTS LTD
Description: The C-10 cabinet is designed for use with compact UV lamps (4 W, such as models UVGL-15 or UVGL-25, or handheld UV lamps (6 W), such as the UVGL-58. T...

Catalog Number: (UVPA95-0253-02)
Supplier: UVP ULTRA VIOLET PRODUCTS LTD
Description: UV viewing cabinet, With two UV tubes, Chromato-Vue®, C-75, 2×15 W, 254/365 nm
UOM: 1 * 1 items


Catalog Number: (UVPA95-0022-02)
Supplier: UVP ULTRA VIOLET PRODUCTS LTD
Description: UV viewing cabinet, Chromato-Vue®, C-71, 4×15 W, 365 nm
UOM: 1 * 1 items


Catalog Number: (UVPA95-0257-02)
Supplier: UVP ULTRA VIOLET PRODUCTS LTD
Description: UV viewing cabinet, Chromato-Vue®, C-65, 2×8 W
UOM: 1 * 1 items


Catalog Number: (UVPA95-0020-05)
Supplier: UVP ULTRA VIOLET PRODUCTS LTD
Description: The large, lightweight plastic UV Chromato-Vue® viewing cabinet C-70G is designed for examining fluorescent samples in a fully lit environment.
UOM: 1 * 1 items


Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
Product(s) marked with this symbol are discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.
1 - 5 of 5
no targeter for Bottom