Multichannel tube pumps. REGLO Digital series
Supplier: Ismatec
224-1020EA 2330 EUR
ISMCISM832C ISMCISM834C ISMCISM831C ISMCISM596D ISMCISM597D ISMCISM597F
Multichannel tube pumps. REGLO Digital series
REGLO Digital series. Compact, space saving peristaltic pumps with metering function. Microprocessor. High reproducibility.
  • Two or four channels
  • Available with 6, 8, or 12 rollers (very low pulsation)
  • Click´n´go cassettes with automatic contact pressure
  • Speed setting: min-1, increment 0,1 min-1 via membrane keypad with LED display
  • For three stop tubing
  • Interfaces: Controllable via PC, analogue: Speed output, speed control (0 to 5 or 0 to 10 V, 0 to 20 or 4 to 20 mA), speed output (2-channel: 0 to 8 kHz/4-channel: 0 to 5 kHz), start/stop, rotation direction, autostart