You Searched For: Solid Samplers
Solid samplers collect portions of bulk goods and are suitable for use with a range of sample substrates from fine powder to coarse grain. Taking cross-sectional samples from different sample depths, the tools are manufactured for easy cleaning and sterilizing in between uses. Conical stainless steel models are ideal for cutting out samples from soft, paste-like substances such as cheese, butter, clay, loam, soap, or wax. Solid samplers are designed specifically to extract even from difficult locations.
68 results were found
Sort Results
List View
Easy View