Combined incubators with variable speed orbital shakers that are designed to provide controlled and uniform conditions for biological growth. The smoked acrylic side panels and door allow visibility of vessels during incubation; forced air circulation ensures uniform heating. SI600C has double glazed viewing windows to prevent condensation when the unit is set at sub-ambient temperatures. The accessory racks can be quickly locked into place or removed from the platform without the need for tools via the magnalock system. The stainless steel tube accessory racks can also adjust the angle of the tubes up to 30°, which increases the surface area of samples. The platform accommodates most sizes of flasks, up to 1 litre on the SI500, typically the SI500 platform accepts 12×250, 9×500, or 4×1000 ml. The SI600/SI600C can accommodate 15×500l, 8×1000, or 6×2000 ml Erlenmeyer flasks. Four securing bars can be adjusted vertically or horizontally to suit. USB communications port allows unit to be connected to a PC for long term temperature monitoring.
- Retractable platform allows easy access to samples at the back
- Digital speed and temperature control via easy to read LED control panel
- All units control up to 60 °C, the SI600C when combined with the SRC4 recirculating cooler can be used down to 15 °C below ambient (min. 5 °C)
- Integral timer which can be set from 1 s to 9 days, after which shaking will stop and unit remain at set temperature to protect samples
- Adjustable safety temperature cut-out provides extra protection for samples and shaking automatically stops when door is opened for improved user safety
- BioCote® antimicrobial protection
Certifications: IP protection class according to DIN EN 60529: IP 22
Delivery information: Supplied with retractable platform with clamping system, but without accessory racks which must be ordered separately.
Caution: * SI600C requires separate recirculating cooler