You Searched For: Vacuum Gauges


11  results were found

SearchResultCount:"11"

Sort Results

List View Easy View

Rate These Search Results

Supplier: VACUUBRAND GMBH + CO
Description: VACUU·VIEW vacuum gauge are chemically resistant ceramic diaphragm sensor for measurement in the rough vacuum range and it provides gas-independent pr...

Safer Human and Environmental Health

Catalog Number: (181-1031)
Supplier: VACUUBRAND GMBH + CO
Description: Versatile, accurate and robust vacuum gauge. Outstanding measurement accuracy thanks to its capacitive, anti-corrosive gauge head with excellent long-...
UOM: 1 * 1 items

Safer Human and Environmental Health


Catalog Number: (181-0624)
Supplier: VACUUBRAND GMBH + CO
Description: The DVR 2pro is a fully electronic, versatile vacuum gauge for the measuring range from atmospheric pressure to 1 mbar.
UOM: 1 * 1 items


Supplier: VACUUBRAND GMBH + CO
Description: The DCP 3000 is an outstanding vacuum gauge with large illuminated display. The brand new VSP 3000 vacuum sensor is based on thermal conductivity (Pir...

Supplier: EDWARDS HIGH VACUUM (METTLER)
Description: The active gauge displays are a range of simple, low cost units for the measurement and control of vacuum from atmosphere to 10-9 mbar.

Supplier: EDWARDS HIGH VACUUM (METTLER)
Description: Model APG.
These gauges have a measuring range from atmosphere to 1×10⁻⁴ mbar and combine the gauge head and controller in one compact active un...

Catalog Number: (EDWAD39591500)
Supplier: EDWARDS HIGH VACUUM (METTLER)
Description: This Active Digital Controller (ADC) is a compact single gauge controller with automatic recognizes compatible Edwards gauges, loads the appropriate l...
UOM: 1 * 1 items


Catalog Number: (181-9915)
Supplier: KOEHLER TECHNISCHE PRODUKTEN
Description: Simple brass vacuum/pressure gauge for inclusion in vacuum lines to give coarse vacuum/pressure indication.
UOM: 1 * 1 items


Catalog Number: (NEUB2283-10-1000)
Supplier: NEUBERT VOLUM. GLASSWAERE
Description: It is complete with adapter on core NS 29/32 and suitable for work in the rough vacuum range.
UOM: 1 * 1 items


Catalog Number: (619-0222)
Supplier: EDWARDS HIGH VACUUM (METTLER)
Description: Active Inverted Magnetron gauge is a combined unit of the gauge-head and controller. This gauge operates as a cold cathode ionisation gauge, in which ...
UOM: 1 * 1 items


Catalog Number: (EDWAD39700000)
Supplier: EDWARDS HIGH VACUUM (METTLER)
Description: TIC Instrument controller is a compact control with a large, clear graphical display, an intuitive user interface and serial communications.
UOM: 1 * 1 items


Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
Product(s) marked with this symbol are discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.
1 - 11 of 11
no targeter for Bottom