You Searched For: Water Dispensers

Essential for high traffic public areas, water dispensers provide a steady stream of clean water. The sloped basin feeds liquids into the bottom drain for disposal. With easy-to-operate on and off controlling, the stainless steel devices conserve water and power for maximum efficiency. Available in compact designs, the free standing or wall-tight water dispensers fit any venue to solve the issue of a space lacking a sink and are simple to install.

Essential for high traffic public areas, water dispensers provide a steady stream of clean water. The sloped basin feeds liquids into the bottom drain for disposal. With easy-to-operate on and off controlling, the stainless steel devices conserve water and power for maximum efficiency. Available in compact designs, the free standing or wall-tight water dispensers fit any venue to solve the issue of a space lacking a sink and are simple to install.


4  results were found

SearchResultCount:"4"

Sort Results

List View Easy View

Rate These Search Results

Supplier: Sartorius
Description: Extension tubing set (4 m) for the Arium® Smart Station is designed to provide uncomplicated extension of the connection between the Arium® Smart Stat...

Supplier: Sartorius
Description: The Arium® Smart Station can be optimised to fit easily into any lab environment. Combine the Smart Station with the water system type needed to dispe...

Supplier: Sartorius
Description: The Arium® Smart Station can be optimised to fit easily into any lab environment. Combine the Smart Station with the water system type needed to dispe...

Catalog Number: (474-0035)
Supplier: SCOTSMAN
Description: The SWC14B water cooler has two independent taps, one controlled by a foot pedal the other is hand activated, both have automatic stream regulation. U...
UOM: 1 * 1 items


Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
Product(s) marked with this symbol are discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.
1 - 4 of 4
no targeter for Bottom