ß-Nicotinamide adenine dinucleotide phosphate (NADP-K, oxidized form) ca. 90%

Supplier: Thermo Scientific
Warning

B24450.77EA 47.4 EUR
B24450.77 B24450.02 B24450.88
ß-Nicotinamide adenine dinucleotide phosphate (NADP-K, oxidized form) ca. 90%
ß-Nicotinamide adenine dinucleotide phosphate (NADP-K, oxidized form)
Formula: C₂₁H₂₇KN₇O₁₇P₃
Melting Pt: ∼ 175 °C (dec.)
Storage Temperature: Freezer
MDL Number: MFCD00148915
CAS Number: 68141-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR