Perchloric acid ≥70%, AnalaR NORMAPUR® analytical reagent

Supplier: VWR Chemicals
Danger

101761VEA 0 EUR
101761V
Perchloric acid ≥70%, AnalaR NORMAPUR® analytical reagent
Perchloric acid
Formula: HClO₄
MW: 100,46 g/mol
Boiling Pt: 160…181 °C (1013 hPa)
Melting Pt: –112,2 °C
Density: 1,53…1.768 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011325
CAS Number: 7601-90-3
EINECS: 231-512-4
UN: 1873
ADR: 5.1,I
https://ie.vwr.com/stibo/web/std.lang.all/06/22/16640622.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR