Perchloric acid ≥70%, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis (max. 0.005 ppm Hg), Supelco®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ300

1.00514.1006EA 2230 EUR
1.00514.1006 1.00514.1000
Perchloric acid ≥70%, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis (max. 0.005 ppm Hg), Supelco®
Perchloric acid
Formula: HClO₄
MW: 100,46 g/mol
Boiling Pt: 160…181 °C (1013 hPa)
Melting Pt: –112,2 °C
Density: 1,53…1.768 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011325
CAS Number: 7601-90-3
EINECS: 231-512-4
UN: 1873
ADR: 5.1 (8),I
https://ie.vwr.com/stibo/web/std.lang.all/06/22/16640622.jpg

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -18 °C
Boiling Point 198.7 °C (1013 hPa)
Density 1.68 g/cm3 (20 °C)
pH (H2O, 20 °C) strongly acid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR