Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (97)
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "Acetonitrile"

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"79"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  Merck
Description:   With their high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level, LiChrosolv® solvents are...
Supplier:  Merck
Description:   Acetonitrile with 0.1% (v/v) Formic acid hypergrade for LC-MS LiChrosolv® 1 * 1 l
Supplier:  Merck
Description:   Acetonitrile EMPLURA®
Supplier:  Merck
Description:   Acetonitrile for spectroscopy Uvasol® 1 * 1 l
Supplier:  Merck
Description:   Acetonitrile EMSURE® ACS, Reag. Ph. Eur. analytical reagent
Supplier:  Merck
Description:   Acetonitrile dried (max. 0.005 % H2O) SeccoSolv® 1 * 150 ml
Supplier:  Merck
Description:   Acetonitrile for gas chromatography SupraSolv® 1 * 1 l
Supplier:  Merck
Description:   ACETONITRILE FOR GAS CHROMATOGRAPHY MS SUPRASOLV® 1 * 1 l
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is dicontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.
1 - 8  of 79
  1  2  3  4  5  6  7  8  9  10  Next