Tris(perfluorobutyl)amine , Perfluoro-compound FC-43™

Supplier: Thermo Scientific

Synonyms: Perfluorotri-n-butylaminePerfluorotributylamineTris(nonafluorobutyl)aminePFTBAFC-43HeptacosafluorotributylamineHeptacosafluorotri-n-butylamine

ACRO123771000EA 465 EUR
ACRO123771000 ACRO123770100 ACRO123772500
Tris(perfluorobutyl)amine , Perfluoro-compound FC-43™
Tris(perfluorobutyl)amine
Formula: C₁₂F₂₇N
MW: 671,1 g/mol
Boiling Pt: 155 °C (1013 hPa)
Melting Pt: –52 °C
Density: 1,88 g/cm³
MDL Number: MFCD00000436
CAS Number: 311-89-7
EINECS: 206-223-1

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Boiling point 173 to 181 °C
Density (25 °C) 1.865 to 1.895 g/ml
Additional info distillation range (between 165 and 185 °C): > 97%
Additional info residue: 10 ug/ml
Additional info pour point: < - 40 °C
Additional info dielectric strength: 35 kV
Viscosity 2.3 to 3.3 cSt (at 25 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR