Tris(perfluorobutyl)amine, Supelco®
Supplier: Merck
Synonyms:
Perfluorotri-n-butylamine
, Perfluorotributylamine
, Tris(nonafluorobutyl)amine
, PFTBA
, FC-43
, Heptacosafluorotributylamine
, Heptacosafluorotri-n-butylamine
M-Clarity™ quality level = MQ100
442747-UEA
56.6
EUR
442747-U
Tris(perfluorobutyl)amine, Supelco®
Tris(perfluorobutyl)amine
Formula:
C₁₂F₂₇N MW: 671,1 g/mol Boiling Pt: 155 °C (1013 hPa) Melting Pt: –55 °C Density: 1,883 g/cm³ (20 °C) |
MDL Number:
MFCD00000436 CAS Number: 311-89-7 EINECS: 206-223-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...