2-Bromo-4-(trifluoromethyl)benzenesulphonamide

Supplier: Apollo Scientific

APOSPC5412-5GEA 43.9 EUR
APOSPC5412-5G
2-Bromo-4-(trifluoromethyl)benzenesulphonamide
2-Bromo-4-(trifluoromethyl)benzenesulphonamide
Formula: C₇H₅BrF₃NO₂S
MW: 304,09 g/mol
MDL Number: MFCD03094287
CAS Number: 351003-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR