5-Bromo-2-fluoro-3-methylpyridine 95%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
5-Bromo-2-fluoro-3-picoline
H26131.03EA
107
EUR
H26131.03
5-Bromo-2-fluoro-3-methylpyridine 95%
5-Bromo-2-fluoro-3-methylpyridine
Formula:
C₆H₅BrFN MW: 190,01 g/mol Melting Pt: 60…63 °C Storage Temperature: Ambient |
MDL Number:
MFCD03095091 CAS Number: 29312-98-9 EINECS: None |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...