Glycerine ≥99.0%, Proteomics Grade
Synonyms:
Propane-1,2,3-triol
, Glycerol
, 1,2,3-Propanetriol
M152-1LEA
105
EUR
M152-1L
Glycerine ≥99.0%, Proteomics Grade
Glycerine
Formula:
HOCH₂CH(OH)CH₂OH MW: 92,09 g/mol Boiling Pt: 182 °C Melting Pt: 20 °C Density: 1,26 g/cm³ (20 °C) Flash Pt: 177 °C (open cup) Storage Temperature: Ambient |
MDL Number:
MFCD00004722 CAS Number: 56-81-5 EINECS: 200-289-5 Merck Index: 13,04497 |
Specification Test Results
Arsenic | <0.0003% |
Chloride | 0.001% |
DNase | None |
Heavy metals (as Pb) | <0.0005% |
Protease | None |
Purity (Anhydrous) | 99.0% |
Residue on ignition | 0.01% |
RNase | None |
Specific Gravity Measurement | 1.2615 − 1.2635 |
Sulfate | 0.001% |
Water (KF) | 1.0% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...