n-Docosane, Supelco®

Supplier: Merck

Synonyms: N-docosane

M-Clarity™ quality level = MQ100

43942-5GEA 187 EUR
43942-5G 43942-1G
n-Docosane, Supelco®
n-Docosane
Formula: H₃C(CH₂)₂₀CH₃
MW: 310,61 g/mol
Boiling Pt: 368 °C (1013 hPa)
Melting Pt: 44 °C
Density: 0,7944 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009348
CAS Number: 629-97-0
EINECS: 211-121-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR