Potassium-3-iodophenyltrifluoroborate 96%

Supplier: Thermo Scientific
Warning

H29009.06EA 0 EUR
H29009.06 H29009.03
Potassium-3-iodophenyltrifluoroborate 96%
Potassium 3-iodophenyltrifluoroborate
Formula: C₆H₄BIF₃K
MW: 309,91 g/mol
Melting Pt: 202…204 °C
Storage Temperature: Ambient
MDL Number: MFCD09800739
CAS Number: 1189097-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR