2,2,3,3,3-Pentafluoro-1-propanol
|
Warning
|
Synonyms:
1H,1H-Pentafluoro-1-propanol
, 1H,1H-Pentafluoropropanol-1
, PFPOH (pentafluoropropanol)
HYPETS-65195EA
707
EUR
HYPETS-65195
2,2,3,3,3-Pentafluoro-1-propanol
2,2,3,3,3-Pentafluoro-1-propanol
Formula:
C₃H₃F₅O MW: 150,05 g/mol Boiling Pt: 78…80 °C (1013 hPa) Density: 1,5 g/cm³ (20 °C) |
MDL Number:
MFCD00004673 CAS Number: 422-05-9 EINECS: 207-012-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...