n-Tetradecane
Supplier: LGC Standards PROMOCHEM
|
Danger
|
Synonyms:
Tetradecane
PROOCHE154EA
237
EUR
PROOCHE154
n-Tetradecane
n-Tetradecane
Formula:
H₃C(CH₂)₁₂CH₃ MW: 198,39 g/mol Boiling Pt: 253,7 °C (1013 hPa) Melting Pt: 6 °C Density: 0,7628 g/cm³ (20 °C) Flash Pt: 102 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00008986 CAS Number: 629-59-4 EINECS: 211-096-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...