2-Methylbutyraldehyde 95%
Supplier: Thermo Scientific
|
Danger
|
A19650.22EA
107
EUR
A19650.22
A19650.09
A19650.14
2-Methylbutyraldehyde 95%
2-Methylbutyraldehyde
Purity: 95%
CAS No.: 96-17-3
Molecular Formula: C5H10O
Formula Weight: 86.13
Flash Point: 4°(39°F)
Refractive Index: 1.3919
Boiling Point: 94-96°
Beilstein Reference: 1633540
UN No.: 1989
MDL No.: MFCD00006984
Formula:
C₅H₁₀O MW: 86,13 g/mol Boiling Pt: 94…96 °C Melting Pt: –100 °C Density: 0,806 Flash Pt: 4 °C (39 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006984 CAS Number: 96-17-3 EINECS: 202-485-6 UN: 1989 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...