1,1-Bis(methylthio)-2-nitroethylene
Supplier: Molekula
|
Warning
|
MOLEM13679748EA
0
EUR
MOLEM13679748
1,1-Bis(methylthio)-2-nitroethylene
1,1-Bis(methylthio)-2-nitroethylene
Formula:
C₄H₇NO₂S₂ MW: 165,24 g/mol Melting Pt: 122…125 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00010443 CAS Number: 13623-94-4 EINECS: 237-108-4 UN: 2811 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...