The modular concept of the Cubis® range has been extended to microbalances for highly accurate weighing of microgram quantities. The range meets the most stringent requirements of analytical laboratories and pharmaceutical research, offering the user the highest level of safety in terms of result reliability and standard conformity.
- Choice of draught shield: Filter weighing (DF); Automatic (DM)
- isoCAL – full and configurable management of the microbalance's calibration routines
- Q-Level – warning when your balance is out of level and easy guidance for re-levelling
- GLP-compliant data printout - user-configurable for MSA models
- Q-Com full communications package USB and RS232 with Bluetooth® wireless option
Short measurement times and convenient access to the weighing chamber saves operator time. The motorised 100% glass draught shield means that working with minimum sample sizes is fast and effortless. The intelligent learning capability allows individual adaptation of the opening angle to suit every workflow. With assisted leveling.
Microbalances for filter weighing:
The special DF stainless steel filter draught shield minimises electrostatic effects and is optimised for ultra-precise weighing of filters. Different weighing pan diameters are available for different filter sizes (50 mm as standard - 75 and 90 mm optional).
MSE control unit: Black and white basic backlit LC display
Ideal for straightforward high accuracy weighing
MSA control unit: High resolution colour, tiltable and configurable TFT graphical touch screen display
Advanced Pharma Compliance - password protection allowing definable Administrator and users with appropriate access levels. Full audit trail and Alibi weighing data storage memory
SQ Min – warning prompt when minimum sample weight has not been reached according to the USP
SD card slot and Ethernet for transfer of, not only data, but task configurations and settings from balance to balance or to PC
Q-Guide and Task management options facilitate the easiest possible navigation through the user-interface to manage your daily routines
Ordering information: A range (-0CE suffix) of verified versions for use in legal metrology in the European Economic area is available - please enquire or see website for details.