4-Bromoaniline analytical reagent, Supelco®

Danger


M-Clarity™ quality level = MQ100

1.01599.0025EA 780 EUR
1.01599.0025 1.01599.9999
4-Bromoaniline analytical reagent, Supelco®
4-Bromoaniline
Formula: C₆H₆BrN
MW: 172,02 g/mol
Boiling Pt: 223 °C (1013 hPa)
Melting Pt: 62…64 °C
Density: 1,799 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007822
CAS Number: 106-40-1
EINECS: 203-393-9
UN: 3077
ADR: 9,III
Merck Index: 13,01388

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Melting Point 60 - 64 °C
Molar Mass 172.02 g/mol
Bulk Density 810 kg/m3
Boiling Point 230 - 250 °C (1013 hPa)
Hazard Class 9
Hazard Class Description Miscellaneous hazardous material

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR