6-Fluorosalicylaldehyde

Supplier: Apollo Scientific

Synonyms: 2-Fluoro-6-hydroxybenzaldehyde

APOSPC9248-5GEA 86.4 EUR
APOSPC9248-5G
6-Fluorosalicylaldehyde
6-Fluorosalicylaldehyde
Formula: C₇H₅FO₂
MW: 140,11 g/mol
MDL Number: MFCD01090996
CAS Number: 38226-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR