Methyl indole-6-carboxylate

Supplier: Molekula

Synonyms: Methyl 1H-indole-6-carboxylate

MOLEM49212138EA 0 EUR
MOLEM49212138
Methyl indole-6-carboxylate
Methyl indole-6-carboxylate
Formula: C₁₀H₉NO₂
MW: 175,19 g/mol
MDL Number: MFCD00211063
CAS Number: 50820-65-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR