Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Gramine ≥99%

Supplier: Acros Organics
Danger



Formula: C₁₁H₁₄N₂
MW: 174,25 g/mol
Flash Pt: 167°C
Storage Temperature: Refrigerator
MDL Number: MFCD00005629
CAS Number: 87-52-5
EINECS: 201-749-8
UN: 3259
ADR: 8,III
Specification Test Results
Appearance Beige Powder

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle 500 g ACRO120185000
Glass bottle 10 g ACRO120180100
Glass bottle 25 g ACRO120180250
Glass bottle 100 g ACRO120181000
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Melting point 131°C - 139°C 131°C - 139°C
Flash point 167°C 167°C
CAS No. 87-52-5 87-52-5
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty