2,3-Dimethyl-5-nitroindole ≥97%
Supplier: Thermo Scientific
Synonyms:
2,3-Dimethyl-5-nitro-1H-indole
L07644.06EA
0
EUR
L07644.06
2,3-Dimethyl-5-nitroindole ≥97%
2,3-Dimethyl-5-nitroindole
Formula:
C₁₀H₁₀N₂O₂ MW: 190,2 g/mol Melting Pt: 186…192 °C Storage Temperature: Ambient |
MDL Number:
MFCD00022708 CAS Number: 21296-94-6 EINECS: 244-321-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...