(R)-(+)-1-(2-Naphthyl)ethylamine ≥99%, ee 99%, ChiPros®
Synonyms:
(R)-1-(2-Naphthyl)ethylamine
L17023.03EA
0
EUR
L17023.03
(R)-(+)-1-(2-Naphthyl)ethylamine ≥99%, ee 99%, ChiPros®
(R)-(+)-1-(2-Naphthyl)ethylamine
Formula:
C₁₂H₁₃N MW: 171,24 g/mol Melting Pt: 45…52 °C |
MDL Number:
MFCD00085367 CAS Number: 3906-16-9 ADR: , |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...