Benzaldehyde oxime, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Benzaldoxime
, alpha-benzaldoxime
M-Clarity™ quality level = MQ200
8.20234.0025EA
113
EUR
8.20234.0025
8.20234.9999
Benzaldehyde oxime, Sigma-Aldrich®
Benzaldehyde oxime
Formula:
C₇H₇NO MW: 121,14 g/mol |
MDL Number:
MFCD00002119 CAS Number: 932-90-1 EINECS: 213-261-2 |
Specification Test Results
Solubility | (20 °C) slightly soluble |
Melting Point | 24 - 27 °C |
Molar Mass | 121.14 g/mol |
Density | 1.11 g/cm3 |
Synonyms | Benzaldoxime |
Hill Formula | C7H7NO |
Chemical Formula | C6H5CH=NOH |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...