2-Methoxyphenethylamine

Supplier: Molekula

MOLE30113132-25GEA 0 EUR
MOLE30113132-25G
2-Methoxyphenethylamine
2-Methoxyphenethylamine
Formula: C₉H₁₃NO
MW: 151,21 g/mol
MDL Number: MFCD00008186
CAS Number: 2045-79-6
EINECS: 218-066-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR