Cyclododecanone for synthesis, Sigma-Aldrich®


M-Clarity™ quality level = MQ200

8.02676.9999
Cyclododecanone for synthesis, Sigma-Aldrich®
Cyclododecanone
Formula: C₁₂H₂₂O
MW: 182,31 g/mol
Boiling Pt: 261 °C (1013 hPa)
Melting Pt: 59…61 °C
Density: 0,87 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003722
CAS Number: 830-13-7
EINECS: 212-595-6
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Solubility 0.045 g/l (20 °C)
Melting Point 59 - 61 °C
Molar Mass 182.3 g/mol
Boiling Point 277 °C (1013 hPa)
Vapor Pressure  - 1 hPa (20 °C)
Flash Point 118 °C
Density 0.97 g/cm3 (20 °C)
Ignition Point 230 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR