2-Bromo-6-formylpyridine
Supplier: Molekula
Synonyms:
6-Bromopyridine-2-carboxaldehyde
, 6-Bromopyridine-2-carbaldehyde
, 6-Bromo-2-pyridinecarboxaldehyde
MOLEM62552937EA
0
EUR
MOLEM62552937
2-Bromo-6-formylpyridine
2-Bromo-6-formylpyridine
Formula:
C₆H₄BrNO MW: 186,01 g/mol |
MDL Number:
MFCD02683546 CAS Number: 34160-40-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...