Sodium carbonate, anhydrous ≥99.5% ACS

Supplier: VWR Chemicals
Warning

0585-2.5KGEA 264 EUR
0585-2.5KG 0585-500G 0585-1KG
Sodium carbonate, anhydrous ≥99.5% ACS
Sodium carbonate
Formula: Na₂CO₃
MW: 105,99 g/mol
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 854 °C
Density: 2,53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
EINECS: 207-838-8
REACH: 01-2119485498-19
Merck Index: 14,08596
https://ie.vwr.com/stibo/web/std.lang.all/05/89/16640589.jpg

Order Now

Specification Test Results

Calcium <0.03%
Chloride ≤0.001%
Heavy metals (as Pb) ≤0.0005%
Insolubles ≤0.01%
Iron ≤0.0005%
Loss on heating ≤1%
Magnesium ≤0.005%
Phosphate ≤0.001%
Potassium ≤0.005%
Purity ≥99.5%
Silica ≤0.005%
Sulphur compounds ≤0.003%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR