Potassium iodide, BAKER, Laboratory Reagent, J.T.Baker®

Supplier: Avantor

0228.1000EA 0 EUR
0228.1000
Potassium iodide, BAKER, Laboratory Reagent, J.T.Baker®
Potassium iodide
Formula: Kl
MW: 166 g/mol
Boiling Pt: 1330 °C (1013 hPa)
Melting Pt: 686 °C
Density: 3,13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0
EINECS: 231-659-4
Merck Index: 12,07809

Order Now

Specification Test Results

Assay (KI)(Dried Basis) 99.0 - 101.5%
Alkalinity Passes Test
Heavy Metals (as Pb) Max. 0.001%
Identification Passes Test
Iodate (IO₃) Max. 4 ppm
Loss on Drying at 105 °C Max. 1.0%
Nitrate, Nitrite and Ammonia Passes Test
Thiosulfate and Barium Passes Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR