Distributors for Reaction Vessels(S), BOHLENDER
BOHLD746-16EA 182 EUR
BOHLD746-16
Distributors for Reaction Vessels(S), BOHLENDER
Blue screw cap made of PP with GLS 80 thread, body made of PP with center ground joint and lateral GL-threaded necks.
  • Temperature resistance: from 0 to +110 °C
  • Very good chemical resistance
  • Vacuum: suitable
  • Autoclavable: 121 °C