Sodium sulphate, anhydrous, powder, AnalaR NORMAPUR® analytical reagent

Supplier: VWR Chemicals

Synonyms: Sulphuric acid disodium saltBisodium sulphateDisodium sulphate

102644VEA 0 EUR
102644V
Sodium sulphate, anhydrous, powder, AnalaR NORMAPUR® analytical reagent
Sodium sulphate
Formula: Na₂SO₄
MW: 142,04 g/mol
Boiling Pt: 1429 °C (1013 hPa)
Melting Pt: 888 °C
Density: 2,7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003504
CAS Number: 7757-82-6
EINECS: 231-820-9
Merck Index: 12,08829
https://ie.vwr.com/stibo/web/std.lang.all/06/41/16640641.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR