Magnesium chloride, ultra dry ≥99.9% (metals basis)
Supplier: Thermo Scientific
Warning
|
ampouled under argon
35707.09EA
114
EUR
35707.09
35707.18
Magnesium chloride, ultra dry ≥99.9% (metals basis)
Magnesium chloride
Formula:
MgCl₂ MW: 95,21 g/mol Boiling Pt: 1412 °C (1013 hPa) Melting Pt: 714 °C Density: 2,32 Storage Temperature: Ambient |
MDL Number:
MFCD00011106 CAS Number: 7786-30-3 EINECS: 232-094-6 Merck Index: 13,05684 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...