Safety shoes for ladies, slip-on, Temptation® City Image
Supplier: Honeywell Safety Products
DALL6551135/38EA 116 EUR
DALL6551135/38 DALL6551135/37 DALL6551135/39 DALL6551135/41 DALL6551135/40 DALL6551135/42
Safety shoes for ladies, slip-on, Temptation® City Image
These ladies shoes, with buckled fastenings, are made from water resistant leather, leather lining plus a PU outsole.
  • Leather lining and leather insole provide comfort and durability
  • Flexible, light sole, slip-resistant on different types of surfaces
  • Protective steel toecap (200 J)
Suitable for women working in services and utilities.
Colour: Black
Certifications: EN ISO 20345:2007; S2, SRC