Sodium hydroxide 0.005 mol/l (0.005 N) in methanol, LiChrosolv® for HPLC, Supelco®

Supplier: Merck

Synonyms: Caustic soda

M-Clarity™ quality level = MQ100

4.80621.9010 4.80621.9030
4.80621.9010EA 0 EUR
4.80621.9010 4.80621.9030
Sodium hydroxide 0.005 mol/l (0.005 N) in methanol, LiChrosolv® for HPLC, Supelco®
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 3286
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR