Cut resistant gloves, Resistop Long
Supplier: Honeywell Safety Products
Resistop
DALLRGT15099G/9EA 75.6 EUR
DALLRGT15099G/9 DALLRGT15099G/8 DALLRGT15099G/7
Cut resistant gloves, Resistop Long
Gloves
These gloves with long cuffs are knitted from 73% polyamide and 27% cotton.
  • Knitting technique helps to obtain a glove that conforms snugly to the hand and good dexterity
  • Cotton interior slows down perspiration and increases comfort
  • Relative thickness helps to absorb shocks
  • Non pilling, the polyamide is suitable for precision work
  • Polyamide offers outstanding abrasion and cutting resistance
  • Long cuff helps to protect a large part of the forearm
Ideal for the handling of sharp parts in automotive and mechanical industries. Also suitable for assembling operations and removal of molds up to 100 °C (for 30 seconds maximum).
Certifications: EN 388:2016 (2X4XB), EN 407 (X1XXXX); Cat. II