Electrical resistant gloves, Electrosoft Composite CL 2
Supplier: Honeywell Safety Products
Electrosoft
DALL2092027/10EA 0 EUR
DALL2092027/10 DALL2092027/11 DALL2092027/9
Electrical resistant gloves, Electrosoft Composite CL 2
Gloves
Natural latex insulating gloves with external layer in orange polychloroprene for electrical work with a maximum voltage of 17 500 volts.
  • Ergonomic shape and lightly powdered interior make the glove easy to slip on and off
  • Natural latex base offers high dielectric characteristics
  • Anti-slip finish on fingers
  • Thickness ensures good dexterity while offering protection up to 17500 volts
  • Arc flash rating of 60,3 cal/cm²
Can be used without leather glove.
Certifications: EN 60903-2003, CEI 903-2002; Cat. III